Anti-Progesterone Receptor (phospho S190) Rabbit Monoclonal Antibody [clone: EP1516Y]

Lieferant: Abcam
AB75857-100 AB75857-1ML
ABCAAB75857-100EA 630 CHF
ABCAAB75857-100 ABCAAB75857-1ML
Anti-Progesterone Receptor (phospho S190) Rabbit Monoclonal Antibody [clone: EP1516Y]
Antikörper
Rabbit monoclonal [EP1516Y] to Progesterone Receptor (phospho S190).

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: EP1516Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR