Anti-EIF4EBP1 Rabbit monoclonal antibody unconjugated [clone: EPR654AY]

Lieferant: Abcam
AB75831-100 AB75831-10
ABCAAB75831-100EA 630 CHF
ABCAAB75831-100 ABCAAB75831-10
Anti-EIF4EBP1 Rabbit monoclonal antibody unconjugated [clone: EPR654AY]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: monoclonal
Clone: EPR654AY
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR