Anti-FSTL1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB71548-100EA 516.36 CHF
ABCAAB71548-100
Anti-FSTL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FSTL1 (follistatin-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR