Anti-Caldesmon Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB68878-100EA 488.19 CHF
ABCAAB68878-100
Anti-Caldesmon Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Caldesmon
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse