Anti-FRK Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB64914-100EA 539.83 CHF
ABCAAB64914-100
Anti-FRK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FRK (fyn-related kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR