Anti-SCN10A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB63331-50EA 516.36 CHF
ABCAAB63331-50
Anti-SCN10A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SCN10A (sodium channel, voltage-gated, type X, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat