Anti-LAMP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB62562-100EA 539.83 CHF
ABCAAB62562-100
Anti-LAMP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR