Anti-Lactate Dehydrogenase B/LDH-B Rabbit Monoclonal Antibody [clone: EP1565Y]

Lieferant: Abcam
AB53292-100 AB53292-1ML
ABCAAB53292-100EA 660 CHF
ABCAAB53292-100 ABCAAB53292-1ML
Anti-Lactate Dehydrogenase B/LDH-B Rabbit Monoclonal Antibody [clone: EP1565Y]
Antikörper
Rabbit monoclonal [EP1565Y] to Lactate Dehydrogenase B/LDH-B.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Lactate Dehydrogenase B/LDH-B
Clonality: Monoclonal
Clone: EP1565Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR