Anti-LPO Sheep polyclonal antibody Biotin

Lieferant: Abcam
ABCAAB34634-1EA 0 CHF
ABCAAB34634-1
Anti-LPO Sheep polyclonal antibody Biotin
Antikörper
Type: Primary
Antigen: LPO (lactoperoxidase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR