Anti-alpha smooth muscle Actin+ACTG2 (acetyl E3) Rabbit Monoclonal Antibody [clone: E184]

Lieferant: Abcam
AB32575-100 AB32575-10 AB32575-1ML
ABCAAB32575-100EA 866 CHF
ABCAAB32575-100 ABCAAB32575-10 ABCAAB32575-1ML
Anti-alpha smooth muscle Actin+ACTG2 (acetyl E3) Rabbit Monoclonal Antibody [clone: E184]
Antikörper
Rabbit monoclonal [E184] to alpha smooth muscle Actin (acetyl E3) + ACTG2 (acetyl E3).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha smooth muscle Actin+ACTG2
Clonality: Monoclonal
Clone: E184
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR