Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: SP2]
Lieferant: Abcam
AB27596-7 AB27596-500 AB27596-1 AB27596-100
ABCAAB27596-7EA 575 CHF
ABCAAB27596-7 ABCAAB27596-500 ABCAAB27596-1 ABCAAB27596-100
Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: SP2]
Antikörper
Rabbit monoclonal [SP2] to Progesterone Receptor, prediluted.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: SP2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human