Anti-GLUT1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB15309-500 AB15309-100
ABCAAB15309-500EA 1610 CHF
ABCAAB15309-500 ABCAAB15309-100
Anti-GLUT1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Glucose Transporter GLUT1.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GLUT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR