Anti-STARD10 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB128946-100 ORB128946-200
BIRBORB128946-100EA 0 CHF
BIRBORB128946-100 BIRBORB128946-200
Anti-STARD10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STARD10 (StAR-related lipid transfer (START) domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR