Anti-ZNF24 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB128936-100 ORB128936-200
BIRBORB128936-100EA 0 CHF
BIRBORB128936-100 BIRBORB128936-200
Anti-ZNF24 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF24 (zinc finger protein 24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR