Anti-RAD50 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB128608-100 ORB128608-200
BIRBORB128608-100EA 0 CHF
BIRBORB128608-100 BIRBORB128608-200
Anti-RAD50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD50 (RAD50 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR