Anti-MKNK1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB128365-100 ORB128365-200
BIRBORB128365-100EA 0 CHF
BIRBORB128365-100 BIRBORB128365-200
Anti-MKNK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MKNK1 (MAP kinase interacting serine/threonine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR