Anti-BCAR1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB127553-100 ORB127553-200
BIRBORB127553-100EA 0 CHF
BIRBORB127553-100 BIRBORB127553-200
Anti-BCAR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BCAR1 (breast cancer anti-estrogen resistance 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR