Anti-AICDA Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB101019-100 ORB101019-200
BIRBORB101019-100EA 0 CHF
BIRBORB101019-100 BIRBORB101019-200
Anti-AICDA Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: AICDA (activation-induced cytidine deaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR