Anti-FUS Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB100082-100 ORB100082-20 ORB100082-200 ORB100082-5
BIRBORB100082-100EA 0 CHF
BIRBORB100082-100 BIRBORB100082-20 BIRBORB100082-200 BIRBORB100082-5
Anti-FUS Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: FUS (fused in sarcoma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR