Anti-AR Goat Polyclonal Antibody

Lieferant: Biorbyt
ORB18847-100 ORB18847-20
BIRBORB18847-100EA 0 CHF
BIRBORB18847-100 BIRBORB18847-20
Anti-AR Goat Polyclonal Antibody
Antikörper
ELISA: 1:64000; Western Blot: 2 ?g/ml

Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR