Anti-alpha Interferon 2b Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB13483-100 ORB13483-200
BIRBORB13483-100EA 0 CHF
BIRBORB13483-100 BIRBORB13483-200
Anti-alpha Interferon 2b Rabbit Polyclonal Antibody
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: alpha Interferon 2b
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR