Anti-KRT7 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
ORB13342-100 ORB13342-200
BIRBORB13342-100EA 0 CHF
BIRBORB13342-100 BIRBORB13342-200
Anti-KRT7 Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat