Anti-IgG1 Mouse Monoclonal Antibody (Biotin)
Lieferant: Biorbyt
ORB12191-100 ORB12191-500
BIRBORB12191-100EA 0 CHF
BIRBORB12191-100 BIRBORB12191-500
Anti-IgG1 Mouse Monoclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: IgG1
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now