Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Lieferant: Biorbyt
ORB12187-100 ORB12187-50
BIRBORB12187-100EA 0 CHF
BIRBORB12187-100 BIRBORB12187-50
Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antikörper
Type: Primary
Antigen: IgG1
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR