Anti-IFABP Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB10869-100 ORB10869-200
BIRBORB10869-100EA 0 CHF
BIRBORB10869-100 BIRBORB10869-200
Anti-IFABP Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IFABP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR