Anti-Digoxin Mouse monoclonal antibody
Lieferant: Biorbyt
ORB10540-100 ORB10540-200
BIRBORB10540-100EA 0 CHF
BIRBORB10540-100 BIRBORB10540-200
Anti-Digoxin Mouse monoclonal antibody
Antikörper
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now