Anti-IFNL2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB6199-100 ORB6199-20 ORB6199-200 ORB6199-5
BIRBORB6199-100EA 0 CHF
BIRBORB6199-100 BIRBORB6199-20 BIRBORB6199-200 BIRBORB6199-5
Anti-IFNL2 Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IFNL2 (interferon, lambda 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR