Anti-E3 ubiquitin ligase Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB2769-100 ORB2769-200
BIRBORB2769-100EA 0 CHF
BIRBORB2769-100 BIRBORB2769-200
Anti-E3 ubiquitin ligase Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: E3 ubiquitin ligase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR