Anti-Maltose Binding Protein Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB2088-100 ORB2088-200
BIRBORB2088-100EA 0 CHF
BIRBORB2088-100 BIRBORB2088-200
Anti-Maltose Binding Protein Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: Maltose Binding Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR