Anti-SKIDA1 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB2029-100 ORB2029-200
BIRBORB2029-100EA 0 CHF
BIRBORB2029-100 BIRBORB2029-200
Anti-SKIDA1 Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: SKIDA1 (SKI/DACH domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR