Anti-LCE1A Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB1453-100 ORB1453-200
BIRBORB1453-100EA 0 CHF
BIRBORB1453-100 BIRBORB1453-200
Anti-LCE1A Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: LCE1A (late cornified envelope 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR