Anti-PPY Rabbit Polyclonal Antibody
Lieferant: Biorbyt
ORB1323-100 ORB1323-200
BIRBORB1323-100EA 0 CHF
BIRBORB1323-100 BIRBORB1323-200
Anti-PPY Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now