Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: HI30]
Lieferant: Tonbo Biosciences
25-0459-T100 25-0459-T025
25-0459-T100EA 250 CHF
25-0459-T100 25-0459-T025
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: HI30]
Antikörper
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.
The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: HI30
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now