Anti-CTDSP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8460AEA 405 CHF
ABGEAP8460A
Anti-CTDSP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTDSP2 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR