Anti-TP53 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP3207AEA 405 CHF
ABGEAP3207A
Anti-TP53 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR