Anti-ATG4A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP1808CEA 405 CHF
ABGEAP1808C
Anti-ATG4A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATG4A (autophagy related 4A, cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR