Anti-MSR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP17001BEA 405 CHF
ABGEAP17001B
Anti-MSR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MSR1 (macrophage scavenger receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR