Anti-TDRD7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP10982BEA 405 CHF
ABGEAP10982B
Anti-TDRD7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TDRD7 (tudor domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR