Anti-VEGFC Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAM1100AEA 405 CHF
ABGEAM1100A
Anti-VEGFC Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: VEGFC (vascular endothelial growth factor C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR