Anti-ANKRD34B Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP67951_P050EA 512 CHF
AVIVARP67951_P050
Anti-ANKRD34B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANKRD34B (ankyrin repeat domain 34B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR