Anti-FASTKD3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
AVIVARP67582_P050EA 512 CHF
AVIVARP67582_P050
Anti-FASTKD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FASTKD3 (FAST kinase domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human