Anti-SPIN2A Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
AVIVARP66685_P050EA 512 CHF
AVIVARP66685_P050
Anti-SPIN2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPIN2A (spindlin family, member 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human