Anti-KIR3DL2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP64058_P050EA 512 CHF
AVIVARP64058_P050
Anti-KIR3DL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIR3DL2 (killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR