Anti-MCPH1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP63864_P050EA 512 CHF
AVIVARP63864_P050
Anti-MCPH1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCPH1 (microcephalin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR