Anti-RASGRP1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP63788_P050EA 512 CHF
AVIVARP63788_P050
Anti-RASGRP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RASGRP1 (RAS guanyl releasing protein 1 (calcium and DAG-regulated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR