Anti-GMPR Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP62557_P050EA 512 CHF
AVIVARP62557_P050
Anti-GMPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GMPR (guanosine monophosphate reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR