Anti-RPL23A Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP61966_P050EA 512 CHF
AVIVARP61966_P050
Anti-RPL23A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL23A (ribosomal protein L23a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR