Anti-STX11 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP61880_P050EA 512 CHF
AVIVARP61880_P050
Anti-STX11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STX11 (syntaxin 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR