Anti-PVRL1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP61313_P050EA 512 CHF
AVIVARP61313_P050
Anti-PVRL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PVRL1 (poliovirus receptor-related 1 (herpesvirus entry mediator C))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR