Anti-CYP24A1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP60147_P050EA 512 CHF
AVIVARP60147_P050
Anti-CYP24A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP24A1 (cytochrome P450, family 24, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR